Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 613667 | ||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
SOBP PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
SOBP - sine oculis binding protein homolog | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_018013.3 | Intron | NP_060483.3 | ||||
XM_005267041.4 | Intron | XP_005267098.1 | ||||
XM_005267042.4 | Intron | XP_005267099.1 | ||||
XM_011535920.2 | Intron | XP_011534222.1 | ||||
XM_011535921.2 | Intron | XP_011534223.1 | ||||
XM_011535923.2 | Intron | XP_011534225.1 | ||||
XM_017010991.1 | Intron | XP_016866480.1 | ||||
XM_017010992.1 | Intron | XP_016866481.1 | ||||
XM_017010993.1 | Intron | XP_016866482.1 | ||||
XM_017010994.1 | Intron | XP_016866483.1 |