Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ACTATTTTGTCATAGGCAGGATTTA[C/T]GGCACTTTGCAGGCTGACTTACCAC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
CCDC63 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
CCDC63 - coiled-coil domain containing 63 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001286243.1 | Intron | NP_001273172.1 | ||||
NM_001286244.1 | Intron | NP_001273173.1 | ||||
NM_152591.2 | Intron | NP_689804.1 | ||||
XM_006719262.1 | Intron | XP_006719325.1 | ||||
XM_006719263.2 | Intron | XP_006719326.1 | ||||
XM_011537999.1 | Intron | XP_011536301.1 | ||||
XM_011538001.2 | Intron | XP_011536303.1 |