Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CATAAACTTGTGTAGGCAGGCAGTT[A/G]TAAGAAGAGTAGCATGCTAAGGGGA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 300230 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
CA5B PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
CA5B - carbonic anhydrase 5B | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_007220.3 | Intron | NP_009151.1 | ||||
XM_005274442.4 | Intron | XP_005274499.1 | ||||
XM_017029247.1 | Intron | XP_016884736.1 | ||||
XM_017029248.1 | Intron | XP_016884737.1 | ||||
XM_017029249.1 | Intron | XP_016884738.1 |