Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GTTATATGCCAGGGACTATTCTATA[C/T]GCTTCAAATACATTATTTAACAATC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 604727 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
STK17B PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
STK17B - serine/threonine kinase 17b | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_004226.3 | Intron | NP_004217.1 | ||||
XM_011512168.2 | Intron | XP_011510470.1 | ||||
XM_011512169.1 | Intron | XP_011510471.1 | ||||
XM_011512170.1 | Intron | XP_011510472.1 | ||||
XM_011512171.2 | Intron | XP_011510473.1 |
Set Membership: |
HapMap |