Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TGCTGGGATTTCACACGTGAACTAC[C/T]GTGCCTGGTTTATTATATTTTGGGG
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||
Literature Links: |
C5orf24 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
C5orf24 - chromosome 5 open reading frame 24 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001135586.1 | Intron | NP_001129058.1 | ||||
NM_001300894.1 | Intron | NP_001287823.1 | ||||
NM_152409.3 | Intron | NP_689622.2 | ||||
XM_005271889.3 | Intron | XP_005271946.1 | ||||
XM_017009049.1 | Intron | XP_016864538.1 | ||||
XM_017009050.1 | Intron | XP_016864539.1 |
DDX46 - DEAD-box helicase 46 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |