Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TGAATGCTGATGGATGCTTTTCTTA[C/T]GCGCAATTTCAACACACTATAGTTC
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 607809 MIM: 601448 | |||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
ACAT1 PubMed Links | |||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | ||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
ACAT1 - acetyl-CoA acetyltransferase 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
NPAT - nuclear protein, coactivator of histone transcription | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001321307.1 | 7259 | Intron | NP_001308236.1 | |||
NM_002519.2 | 7259 | Intron | NP_002510.2 | |||
XM_011542854.2 | 7259 | UTR 3 | XP_011541156.1 | |||
XM_017017802.1 | 7259 | UTR 3 | XP_016873291.1 | |||
XM_017017803.1 | 7259 | UTR 3 | XP_016873292.1 |