Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CACTGGGTCCTTGGATGATCCCAAG[C/T]TGAATCATTGCTTGGGAGGCTCATG
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 614452 | |||||||||||||||||||||||||||||||||||||||||
Literature Links: |
ATAD1 PubMed Links | |||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | ||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
ATAD1 - ATPase family, AAA domain containing 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001321967.1 | Intron | NP_001308896.1 | ||||
NM_001321968.1 | Intron | NP_001308897.1 | ||||
NM_001321969.1 | Intron | NP_001308898.1 | ||||
NM_032810.3 | Intron | NP_116199.2 | ||||
XM_005270252.4 | Intron | XP_005270309.1 | ||||
XM_011540302.1 | Intron | XP_011538604.1 | ||||
XM_011540303.2 | Intron | XP_011538605.1 | ||||
XM_017016847.1 | Intron | XP_016872336.1 | ||||
XM_017016848.1 | Intron | XP_016872337.1 | ||||
XM_017016849.1 | Intron | XP_016872338.1 | ||||
XM_017016850.1 | Intron | XP_016872339.1 | ||||
XM_017016851.1 | Intron | XP_016872340.1 |
CFL1P1 - cofilin 1 pseudogene 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |