Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TGTCCAGCCCTGGGGTTTTCCCATT[G/T]GTGGTTAAGAAGAAGAGTAAGACAG
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 605843 | |||||||||||||||||||||||||||||||||||||||||
Literature Links: |
PECR PubMed Links | |||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | ||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
PECR - peroxisomal trans-2-enoyl-CoA reductase | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
TMEM169 - transmembrane protein 169 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001142310.1 | Intron | NP_001135782.1 | ||||
NM_001142311.1 | Intron | NP_001135783.1 | ||||
NM_001142312.1 | Intron | NP_001135784.1 | ||||
NM_138390.3 | Intron | NP_612399.1 | ||||
XM_006712851.1 | Intron | XP_006712914.1 | ||||
XM_011512173.2 | Intron | XP_011510475.1 |