Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CTTTCAAAATACTCTTCTCTCTTTT[C/T]AAAGTCCCTGACAACTTTACTTCCC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||
Literature Links: |
FAM107B PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
FAM107B - family with sequence similarity 107 member B | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001282695.1 | Intron | NP_001269624.1 | ||||
NM_001282696.1 | Intron | NP_001269625.1 | ||||
NM_001282697.1 | Intron | NP_001269626.1 | ||||
NM_001282698.1 | Intron | NP_001269627.1 | ||||
NM_001282699.1 | Intron | NP_001269628.1 | ||||
NM_001282700.1 | Intron | NP_001269629.1 | ||||
NM_001282701.1 | Intron | NP_001269630.1 | ||||
NM_001282702.1 | Intron | NP_001269631.1 | ||||
NM_001282703.1 | Intron | NP_001269632.1 | ||||
NM_001320735.1 | Intron | NP_001307664.1 | ||||
NM_001320736.1 | Intron | NP_001307665.1 | ||||
NM_001320737.1 | Intron | NP_001307666.1 | ||||
NM_001320738.1 | Intron | NP_001307667.1 | ||||
NM_001320739.1 | Intron | NP_001307668.1 | ||||
NM_001320740.1 | Intron | NP_001307669.1 | ||||
NM_001320741.1 | Intron | NP_001307670.1 | ||||
NM_031453.3 | Intron | NP_113641.2 | ||||
XM_005252616.3 | Intron | XP_005252673.1 | ||||
XM_011519735.1 | Intron | XP_011518037.1 | ||||
XM_017016746.1 | Intron | XP_016872235.1 | ||||
XM_017016747.1 | Intron | XP_016872236.1 | ||||
XM_017016748.1 | Intron | XP_016872237.1 | ||||
XM_017016749.1 | Intron | XP_016872238.1 |