Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AATAAACATTGGCTTGCATATTTCA[A/G]ACCCAAAAGAATTCATTGTGAATTT
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 603317 | ||||||||||||||||||||
Literature Links: |
PTPRQ PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
PTPRQ - protein tyrosine phosphatase, receptor type Q | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001145026.1 | Intron | NP_001138498.1 | ||||
XM_017019273.1 | Intron | XP_016874762.1 | ||||
XM_017019274.1 | Intron | XP_016874763.1 | ||||
XM_017019275.1 | Intron | XP_016874764.1 |