Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AACAAACAGCTTCACAGTTTAATAG[C/T]ATCTTATGTATTTGCCTTCCACTCC
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
29 submissions
|
|||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 248610 | |||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
DBT PubMed Links | |||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | ||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
DBT - dihydrolipoamide branched chain transacylase E2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001918.3 | 6354 | UTR 3 | NP_001909.3 | |||
XM_005270545.3 | 6354 | Intron | XP_005270602.1 | |||
XM_017000468.1 | 6354 | Intron | XP_016855957.1 | |||
XM_017000469.1 | 6354 | Intron | XP_016855958.1 |
Set Membership: |
HapMap |