Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GAGTCTAATGCCACTGCTGATCCAA[C/T]AGGAGGAGGAGCTCAGGTGGTAATC
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 603454 | |||||||||||||||||||||||||||||||||||||||||
Literature Links: |
CRADD PubMed Links | |||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | ||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
CRADD - CASP2 and RIPK1 domain containing adaptor with death domain | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001320099.1 | Intron | NP_001307028.1 | ||||
NM_001320100.1 | Intron | NP_001307029.1 | ||||
NM_001320101.1 | Intron | NP_001307030.1 | ||||
NM_003805.4 | Intron | NP_003796.1 | ||||
XM_017020142.1 | Intron | XP_016875631.1 | ||||
XM_017020143.1 | Intron | XP_016875632.1 | ||||
XM_017020144.1 | Intron | XP_016875633.1 | ||||
XM_017020145.1 | Intron | XP_016875634.1 |