Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TAAAGGAATCCAAGGAACATATCCT[C/T]TGAGAAATAACATCTAAATACTATG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 176851 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
PCMT1 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
PCMT1 - protein-L-isoaspartate (D-aspartate) O-methyltransferase | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001252049.1 | Intron | NP_001238978.1 | ||||
NM_001252050.1 | Intron | NP_001238979.1 | ||||
NM_001252051.1 | Intron | NP_001238980.1 | ||||
NM_001252052.1 | Intron | NP_001238981.1 | ||||
NM_001252053.1 | Intron | NP_001238982.1 | ||||
NM_005389.2 | Intron | NP_005380.2 | ||||
XM_011535868.2 | Intron | XP_011534170.1 |
Set Membership: |
HapMap |