Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CCTAGAAAGTCACTGAACACACCAC[A/G]CAGACAATATAGTCACCATGCAGCA
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
1 submissions
|
||||||||||||||||||||
Phenotype: |
MIM: 608042 | ||||||||||||||||||||
Literature Links: |
SYTL1 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
SYTL1 - synaptotagmin like 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001193308.1 | Intron | NP_001180237.1 | ||||
NM_032872.2 | Intron | NP_116261.1 | ||||
XM_005246022.3 | Intron | XP_005246079.1 | ||||
XM_006710990.1 | Intron | XP_006711053.1 | ||||
XM_006710991.1 | Intron | XP_006711054.1 |
TMEM222 - transmembrane protein 222 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |