Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GATTACCCGGAAAAGCCCTGGCAGG[G/T]AAAGAGAGGACCCGCGCAGGCTGGG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 615138 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
GALNTL6 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
GALNTL6 - polypeptide N-acetylgalactosaminyltransferase-like 6 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001034845.2 | 540 | Intron | NP_001030017.2 | |||
XM_011531993.2 | 540 | Intron | XP_011530295.1 | |||
XM_011531994.1 | 540 | Intron | XP_011530296.1 | |||
XM_011531995.1 | 540 | Intron | XP_011530297.1 | |||
XM_011531996.1 | 540 | Intron | XP_011530298.1 | |||
XM_011531997.1 | 540 | Intron | XP_011530299.1 | |||
XM_017008243.1 | 540 | UTR 5 | XP_016863732.1 | |||
XM_017008244.1 | 540 | Intron | XP_016863733.1 |
LOC441052 - uncharacterized LOC441052 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |