Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CTGGAGCTGCTCACTCGGCCCCGAG[A/C]CCCTTCTCTGACAGTTTACACAAGA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 612078 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
ZNF469 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
ZNF469 - zinc finger protein 469 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001127464.2 | 1291 | Missense Mutation | AGA,AGC | R,S 366 | NP_001120936.2 | |
XM_017023784.1 | 1291 | Missense Mutation | AGA,AGC | R,S 366 | XP_016879273.1 | |
XM_017023785.1 | 1291 | Missense Mutation | AGA,AGC | R,S 366 | XP_016879274.1 |