Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CACTCAGCTGTGTCCATTCTTTATT[A/G]GTTTCCGTAACCATTAAATGTCAGC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
32 submissions
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 600997 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
EPHB2 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
EPHB2 - EPH receptor B2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001309192.1 | Intron | NP_001296121.1 | ||||
NM_001309193.1 | Intron | NP_001296122.1 | ||||
NM_004442.7 | Intron | NP_004433.2 | ||||
NM_017449.4 | Intron | NP_059145.2 | ||||
XM_006710441.3 | Intron | XP_006710504.1 | ||||
XM_006710442.3 | Intron | XP_006710505.1 |
MIR4684 - microRNA 4684 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
Set Membership: |
HapMap |