Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TTTGGGTGACTCACTCTGCCAGGAG[C/T]AGTGTGGATCAAGGACACATTTGTC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 607988 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
SPOCK2 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
SPOCK2 - sparc/osteonectin, cwcv and kazal-like domains proteoglycan (testican) 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001134434.1 | 1977 | Intron | NP_001127906.1 | |||
NM_001244950.1 | 1977 | UTR 3 | NP_001231879.1 | |||
NM_014767.2 | 1977 | UTR 3 | NP_055582.1 | |||
XM_011540404.2 | 1977 | UTR 3 | XP_011538706.1 | |||
XM_017016985.1 | 1977 | Intron | XP_016872474.1 |
Set Membership: |
HapMap |