Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CTCCCTATGGATGTCAGATTGTATC[G/A]TGTCACTCCCCTTCATAAAACCCTT
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 615549 | |||||||||||||||||||||||||||||||||||||||||
Literature Links: |
ARMC5 PubMed Links | |||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | ||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
ARMC5 - armadillo repeat containing 5 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001105247.1 | Intron | NP_001098717.1 | ||||
NM_001288767.1 | Intron | NP_001275696.1 | ||||
NM_001301820.1 | Intron | NP_001288749.1 | ||||
NM_024742.2 | Intron | NP_079018.1 | ||||
XM_006721091.2 | Intron | XP_006721154.1 |