Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ATTTGTGTTTCAGAATTAGAACTAC[A/G]ATACAATAAAATAAAGCCACGCACA
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 609665 MIM: 605015 | |||||||||||||||||||||||||||||||||||||||||
Literature Links: |
NLRP14 PubMed Links | |||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | ||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
NLRP14 - NLR family pyrin domain containing 14 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_176822.3 | Intron | NP_789792.1 | ||||
XM_011520044.1 | Intron | XP_011518346.1 |
ZNF214 - zinc finger protein 214 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |