Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AAGATTTTTACTTGTTTCATTCTCA[A/G]AAAGGCTTGGACAACTATTTGTAGG
Species: |
Human | |||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||
Phenotype: |
MIM: 616121 | |||||||||||||||||||||||
Literature Links: |
GVINP1 PubMed Links | |||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR - Not Available | Japanese - Not Available | JPT (Japanese)
|
|||
EUR - Not Available | |||||
AMR - Not Available |
GVINP1 - GTPase, very large interferon inducible pseudogene 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |