Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 608921 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
FAM64A PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
FAM64A - family with sequence similarity 64 member A | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001195228.1 | 6769 | Intron | NP_001182157.1 | |||
NM_019013.2 | 6769 | Intron | NP_061886.2 | |||
XM_011523944.2 | 6769 | Intron | XP_011522246.1 | |||
XM_017024778.1 | 6769 | Intron | XP_016880267.1 |
PITPNM3 - PITPNM family member 3 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001165966.1 | 6769 | UTR 3 | NP_001159438.1 | |||
NM_031220.3 | 6769 | UTR 3 | NP_112497.2 | |||
XM_011524015.2 | 6769 | Intron | XP_011522317.1 | |||
XM_011524016.2 | 6769 | Intron | XP_011522318.1 | |||
XM_011524017.2 | 6769 | Intron | XP_011522319.1 |
Set Membership: |
HapMap |