Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TGTAACTATAAATCACATTGAGTAA[A/G]AAAAGTACATTACTATTTGTTAATG
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 603541 | ||||||||||||||||||||
Literature Links: |
CCDC38 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
CCDC38 - coiled-coil domain containing 38 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_182496.2 | Intron | NP_872302.2 | ||||
XM_006719229.2 | Intron | XP_006719292.1 | ||||
XM_011537883.2 | Intron | XP_011536185.1 | ||||
XM_011537884.2 | Intron | XP_011536186.1 | ||||
XM_011537886.2 | Intron | XP_011536188.1 | ||||
XM_011537887.2 | Intron | XP_011536189.1 | ||||
XM_011537888.2 | Intron | XP_011536190.1 | ||||
XM_011537889.1 | Intron | XP_011536191.1 |
SNRPF - small nuclear ribonucleoprotein polypeptide F | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |