Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ACTGCTTACCACTTAATTTGTATAG[C/T]CTAGAATAACATAGAATGTTAGTGC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
METTL4 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
METTL4 - methyltransferase like 4 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001308401.1 | 2045 | UTR 3 | NP_001295330.1 | |||
NM_022840.4 | 2045 | UTR 3 | NP_073751.3 | |||
XM_005258132.3 | 2045 | UTR 3 | XP_005258189.1 | |||
XM_005258133.2 | 2045 | UTR 3 | XP_005258190.1 | |||
XM_011525730.2 | 2045 | UTR 3 | XP_011524032.1 |
Set Membership: |
HapMap |