Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TCTACCTGGACCAGCCCAGGAGCCT[A/C]CTTGTTAATCTTGCAGCTTCCACTT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
CCDC85A PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
CCDC85A - coiled-coil domain containing 85A | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001080433.1 | Intron | NP_001073902.1 | ||||
XM_005264125.1 | Intron | XP_005264182.1 |
LOC100129434 - uncharacterized LOC100129434 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
LOC107985795 - uncharacterized LOC107985795 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |