Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AAGTGCTGGGATTATAGGCAACAAC[A/T]TAAATTTATTTTCTCACAATTCTGG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 607617 | ||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
LOC100507250 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
LOC100507250 - uncharacterized LOC100507250 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
NUP107 - nucleoporin 107 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
XM_005269037.3 | Intron | XP_005269094.1 | ||||
XM_011538576.1 | Intron | XP_011536878.1 |
Set Membership: |
HapMap |