Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GATATATAACAGCAACATAGTCTTA[C/T]AGCAATTAATAAATTCCTACAAAGA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 611527 MIM: 611789 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
SLC9B1 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
SLC9B1 - solute carrier family 9 member B1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
SLC9B2 - solute carrier family 9 member B2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001300754.1 | Intron | NP_001287683.1 | ||||
NM_001300756.1 | Intron | NP_001287685.1 | ||||
NM_178833.5 | Intron | NP_849155.2 | ||||
XM_006714085.2 | Intron | XP_006714148.1 | ||||
XM_006714086.3 | Intron | XP_006714149.1 |