Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AAATATGGAAGCAAGGAAAAAGCTG[C/T]CCCTAATCCTGTCACCCTAAGAGAG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 602508 | ||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
PAFAH1B2 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
PAFAH1B2 - platelet activating factor acetylhydrolase 1b catalytic subunit 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001184746.1 | Intron | NP_001171675.1 | ||||
NM_001184747.1 | Intron | NP_001171676.1 | ||||
NM_001184748.1 | Intron | NP_001171677.1 | ||||
NM_001309431.1 | Intron | NP_001296360.1 | ||||
NM_002572.3 | Intron | NP_002563.1 | ||||
XM_017017840.1 | Intron | XP_016873329.1 |
Set Membership: |
HapMap |