Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TTGGAAGTTAGAGAAGTTGAATTGA[A/G]CCATTCCTGGTTTGGGAGCAAAAAG
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 600020 | ||||||||||||||||||||
Literature Links: |
MXI1 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
MXI1 - MAX interactor 1, dimerization protein | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001008541.1 | Intron | NP_001008541.1 | ||||
NM_005962.4 | Intron | NP_005953.4 | ||||
NM_130439.3 | Intron | NP_569157.2 |