Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ATGGAGGGGAGGAGATAAATCAGGT[G/T]GGAGGTAGATGCCATTCAAAAAAAG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 610734 MIM: 613597 MIM: 616388 | ||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
ANKRD2 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | ||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese)
|
||||||
EUR
|
||||||||
AMR
|
ANKRD2 - ankyrin repeat domain 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001129981.2 | Intron | NP_001123453.1 | ||||
NM_001291218.1 | Intron | NP_001278147.1 | ||||
NM_001291219.1 | Intron | NP_001278148.1 | ||||
NM_020349.3 | Intron | NP_065082.2 |
HOGA1 - 4-hydroxy-2-oxoglutarate aldolase 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
UBTD1 - ubiquitin domain containing 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |