Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TCTTTATCCAACTTGCCATAATGTG[C/G]CTTTTAAGTGGGGCATTTAGCCCAT
Species: |
Human | ||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||
Phenotype: |
MIM: 616356 | ||||||||||||||||||||||||||
Literature Links: |
MIR31HG PubMed Links | ||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS - Not Available | Chinese - Not Available | JPT (Japanese)
|
|||
AFR - Not Available | Japanese - Not Available | CHB (Han Chinese)
|
|||
EUR - Not Available | |||||
AMR - Not Available |
MIR31HG - MIR31 host gene | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |