Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GAGGGGTACATTCAATAATAACAAA[A/G]AGCCTTCCAGTTCTAAGTATTTGTG
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 605485 | ||||||||||||||||||||
Literature Links: |
VPS41 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
VPS41 - VPS41, HOPS complex subunit | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_014396.3 | 4298 | UTR 3 | NP_055211.2 | |||
NM_080631.3 | 4298 | UTR 3 | NP_542198.2 | |||
XM_017011988.1 | 4298 | Intron | XP_016867477.1 |