Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TGTTGTTACACAAACTATAGTGTGA[A/C]ACATATTAGTATTTAAAAAACAAAA
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 603290 | |||||||||||||||||||||||||||||||||||||||||
Literature Links: |
SHANK2 PubMed Links | |||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | ||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
SHANK2 - SH3 and multiple ankyrin repeat domains 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_012309.4 | 10148 | UTR 3 | NP_036441.2 | |||
NM_133266.4 | 10148 | UTR 3 | NP_573573.2 | |||
XM_005277932.3 | 10148 | UTR 3 | XP_005277989.1 | |||
XM_017017387.1 | 10148 | UTR 3 | XP_016872876.1 | |||
XM_017017388.1 | 10148 | UTR 3 | XP_016872877.1 | |||
XM_017017389.1 | 10148 | UTR 3 | XP_016872878.1 | |||
XM_017017390.1 | 10148 | UTR 3 | XP_016872879.1 |