Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CCCTCAGCCACAAAGTTCTTTTCCT[A/G]AAACCATAAACACTCTCCATTGTCT
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 604579 | |||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
FZD4 PubMed Links | |||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | ||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
FZD4 - frizzled class receptor 4 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_012193.3 | 4587 | UTR 3 | NP_036325.2 |
LOC100506368 - uncharacterized LOC100506368 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
PRSS23 - protease, serine 23 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001293178.1 | 4587 | Intron | NP_001280107.1 | |||
NM_001293179.1 | 4587 | Intron | NP_001280108.1 | |||
NM_001293180.1 | 4587 | Intron | NP_001280109.1 | |||
NM_007173.5 | 4587 | Intron | NP_009104.2 |
Set Membership: |
HapMap |