Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GGCAGTAAGGAGTTCCCACGTTTGC[C/T]CCGCTTCAGTGTCGTGGACTGGCAA
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||
Phenotype: |
||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
CCEPR PubMed Links | |||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | ||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
CCEPR - cervical carcinoma expressed PCNA regulatory lncRNA | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
PHYHIPL - phytanoyl-CoA 2-hydroxylase interacting protein like | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001143774.1 | 1209 | Intron | NP_001137246.1 | |||
NM_032439.3 | 1209 | Intron | NP_115815.2 | |||
XM_011540275.2 | 1209 | Intron | XP_011538577.1 | |||
XM_011540276.2 | 1209 | UTR 5 | XP_011538578.1 | |||
XM_017016782.1 | 1209 | Intron | XP_016872271.1 | |||
XM_017016783.1 | 1209 | UTR 5 | XP_016872272.1 |
Set Membership: |
HapMap |