Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GTTGAGTGATTTCCATTGTGCTTTT[C/T]CTGTATGGTACATGAACCCTCAAAT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 609202 | ||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
SCGN PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | ||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese)
|
||||||
EUR
|
||||||||
AMR
|
SCGN - secretagogin, EF-hand calcium binding protein | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_006998.3 | Intron | NP_008929.2 | ||||
XM_017010181.1 | Intron | XP_016865670.1 |
Set Membership: |
HapMap |