Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CTCCAGTTGACAACCACTCTATTAA[G/T]GCACTCCCTAATCCTTGGATTATTT
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
1 submissions
|
||||||||||||||||||||
Phenotype: |
MIM: 612866 | ||||||||||||||||||||
Literature Links: |
ALG14 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
ALG14 - ALG14, UDP-N-acetylglucosaminyltransferase subunit | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001305242.1 | Intron | NP_001292171.1 | ||||
NM_144988.3 | Intron | NP_659425.1 | ||||
XM_005270582.3 | Intron | XP_005270639.1 | ||||
XM_011540897.2 | Intron | XP_011539199.1 |