Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TGTGCTTCAATACACAGAGCATTCT[C/T]TGGAAATCCCTACGACACAGGCTTA
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||
Literature Links: |
LOC284009 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
LOC284009 - uncharacterized LOC284009 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
METTL16 - methyltransferase like 16 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_024086.3 | 4346 | UTR 3 | NP_076991.3 | |||
XM_005256801.2 | 4346 | UTR 3 | XP_005256858.1 | |||
XM_017025045.1 | 4346 | UTR 3 | XP_016880534.1 | |||
XM_017025046.1 | 4346 | UTR 3 | XP_016880535.1 |