Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 603032 | ||||||||||||||||||||
Literature Links: |
SNN PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
SNN - stannin | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_003498.5 | 2544 | Intron | NP_003489.1 | |||
XM_017023741.1 | 2544 | Intron | XP_016879230.1 | |||
XM_017023742.1 | 2544 | Intron | XP_016879231.1 |
TXNDC11 - thioredoxin domain containing 11 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001303447.1 | 2544 | Silent Mutation | CAC,CAT | H,H 880 | NP_001290376.1 | |
NM_001324022.1 | 2544 | Silent Mutation | CAC,CAT | H,H 639 | NP_001310951.1 | |
NM_001324024.1 | 2544 | Silent Mutation | CAC,CAT | H,H 401 | NP_001310953.1 | |
NM_001324025.1 | 2544 | Silent Mutation | CAC,CAT | H,H 401 | NP_001310954.1 | |
NM_015914.6 | 2544 | Silent Mutation | CAC,CAT | H,H 853 | NP_056998.4 | |
XM_011522515.2 | 2544 | Silent Mutation | CAC,CAT | H,H 893 | XP_011520817.1 | |
XM_011522516.2 | 2544 | Silent Mutation | CAC,CAT | H,H 679 | XP_011520818.1 | |
XM_017023268.1 | 2544 | Silent Mutation | CAC,CAT | H,H 892 | XP_016878757.1 |