Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 615261 | ||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
METTL22 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | ||||||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | ||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
METTL22 - methyltransferase like 22 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_024109.3 | 13 | UTR 5 | NP_077014.3 | |||
XM_005255570.3 | 13 | UTR 5 | XP_005255627.1 | |||
XM_005255571.2 | 13 | UTR 5 | XP_005255628.1 | |||
XM_011522657.2 | 13 | UTR 5 | XP_011520959.1 | |||
XM_017023659.1 | 13 | UTR 5 | XP_016879148.1 | |||
XM_017023660.1 | 13 | Intron | XP_016879149.1 | |||
XM_017023661.1 | 13 | UTR 5 | XP_016879150.1 | |||
XM_017023662.1 | 13 | Intron | XP_016879151.1 |
Set Membership: |
HapMap |