Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AGAAGTAATTAACTCTCAACCATTT[C/T]GTTCAATTGATATTGAGCTCTTTCT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
34 submissions
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 179755 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
PRCC PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
PRCC - papillary renal cell carcinoma (translocation-associated) | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_005973.4 | Intron | NP_005964.3 | ||||
XM_005245313.2 | Intron | XP_005245370.1 | ||||
XM_005245314.1 | Intron | XP_005245371.1 | ||||
XM_011509726.1 | Intron | XP_011508028.1 |