Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | |||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||
Phenotype: |
MIM: 136537 MIM: 136538 | |||||||||||||||||||||||
Literature Links: |
FPR1 PubMed Links | |||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS - Not Available | African American - Not Available | YRI (Yoruba)
|
|||
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR - Not Available | |||||
AMR - Not Available |
FPR1 - formyl peptide receptor 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
FPR2 - formyl peptide receptor 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001005738.1 | 270 | Intron | NP_001005738.1 | |||
NM_001462.3 | 270 | Intron | NP_001453.1 | |||
XM_006723120.3 | 270 | UTR 5 | XP_006723183.1 | |||
XM_017026542.1 | 270 | UTR 5 | XP_016882031.1 |
Set Membership: |
HapMap |