Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AAACATTTTGCAGTTTACAAAACTA[C/T]ATTTAACATATAGCGATTTGTCTTT
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
39 submissions
|
|||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 613484 | |||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
FLJ37453 PubMed Links | |||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | ||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
FLJ37453 - uncharacterized LOC729614 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
SPEN - spen family transcriptional repressor | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_015001.2 | Intron | NP_055816.2 |
Set Membership: |
HapMap |