Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TGTGTAAGCCACCATGCCCGGCCCA[A/G]AAGTGCTAACTTTTAATGTAGTCAT
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 615753 | ||||||||||||||||||||
Literature Links: |
POM121 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
POM121 - POM121 transmembrane nucleoporin | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001257190.2 | Intron | NP_001244119.1 | ||||
NM_172020.4 | Intron | NP_742017.1 | ||||
XM_005250726.3 | Intron | XP_005250783.1 | ||||
XM_005250727.3 | Intron | XP_005250784.1 | ||||
XM_006716196.3 | Intron | XP_006716259.1 | ||||
XM_011516730.1 | Intron | XP_011515032.1 | ||||
XM_017012853.1 | Intron | XP_016868342.1 | ||||
XM_017012854.1 | Intron | XP_016868343.1 |