Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GGTAAACATGTGCCATGATGGTTTG[C/T]TGCACAGAACAACCCATCACCTAGG
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 123803 | ||||||||||||||||||||
Literature Links: |
ATF1 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
ATF1 - activating transcription factor 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_005171.4 | Intron | NP_005162.1 | ||||
XM_011538386.2 | Intron | XP_011536688.1 | ||||
XM_011538387.2 | Intron | XP_011536689.1 | ||||
XM_011538388.2 | Intron | XP_011536690.1 | ||||
XM_017019331.1 | Intron | XP_016874820.1 | ||||
XM_017019332.1 | Intron | XP_016874821.1 | ||||
XM_017019333.1 | Intron | XP_016874822.1 | ||||
XM_017019334.1 | Intron | XP_016874823.1 | ||||
XM_017019335.1 | Intron | XP_016874824.1 | ||||
XM_017019336.1 | Intron | XP_016874825.1 |