Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TAGCATTAGGATCGTTTTAGACTTA[C/T]AGAAGCATTGCATAGACAGCACAGA
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||
Phenotype: |
||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
MIR3142 PubMed Links | |||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | ||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
MIR3142 - microRNA 3142 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
MIR3142HG - MIR3142 host gene | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |