Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CGATCCTGCCCATTTGGCCCTGCCC[C/T]GATTATTGCTGCCCTTGGCATCTGG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 604411 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
INCENP PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
INCENP - inner centromere protein | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001040694.1 | Intron | NP_001035784.1 | ||||
NM_020238.2 | Intron | NP_064623.2 | ||||
XM_006718533.2 | Intron | XP_006718596.1 | ||||
XM_011544995.2 | Intron | XP_011543297.1 | ||||
XM_011544996.2 | Intron | XP_011543298.1 | ||||
XM_011544997.2 | Intron | XP_011543299.1 | ||||
XM_011544998.2 | Intron | XP_011543300.1 |