Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TGCAAATGTATAAAATTACAATTGA[C/T]TTCTGTATGTTGACCATGTATCCTG
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 615126 MIM: 109635 MIM: 606780 | |||||||||||||||||||||||||||||||||||||||||
Literature Links: |
ANKRD13D PubMed Links | |||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | ||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
ANKRD13D - ankyrin repeat domain 13D | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_207354.2 | Intron | NP_997237.2 | ||||
XM_011544985.1 | Intron | XP_011543287.1 | ||||
XM_011544986.1 | Intron | XP_011543288.1 | ||||
XM_011544987.2 | Intron | XP_011543289.1 | ||||
XM_011544988.2 | Intron | XP_011543290.1 |
GRK2 - G protein-coupled receptor kinase 2 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
SSH3 - slingshot protein phosphatase 3 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |