Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CTTAATTATGTTCAAAAATCCTATT[C/T]TAACTGCTTGAGCTAGGATTGTGCC
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 608016 | ||||||||||||||||||||
Literature Links: |
COA4 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
COA4 - cytochrome c oxidase assembly factor 4 homolog | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
PAAF1 - proteasomal ATPase associated factor 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001267803.1 | Intron | NP_001254732.1 | ||||
NM_001267804.1 | Intron | NP_001254733.1 | ||||
NM_001267805.1 | Intron | NP_001254734.1 | ||||
NM_001267806.1 | Intron | NP_001254735.1 | ||||
NM_025155.2 | Intron | NP_079431.1 | ||||
XM_005274308.2 | Intron | XP_005274365.2 | ||||
XM_005274309.1 | Intron | XP_005274366.1 | ||||
XM_017018357.1 | Intron | XP_016873846.1 | ||||
XM_017018358.1 | Intron | XP_016873847.1 | ||||
XM_017018359.1 | Intron | XP_016873848.1 |