Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TTGGGTTTCTTTTTCAAATCTGGCC[A/G]GTCTATGTCATGTTTCAGCTTACTT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 604987 | ||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
CLEC5A PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | ||||||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | ||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
CLEC5A - C-type lectin domain family 5 member A | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
OR9A4 - olfactory receptor family 9 subfamily A member 4 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001001656.1 | 477 | Silent Mutation | CCA,CCG | P,P 159 | NP_001001656.1 |
Set Membership: |
HapMap |